accessibility menu, dialog, popup

UserName

HNSOOOHOHSOOOHOH2ONa(E/Z)

CAS RN 304655-82-1

IUPAC Name: 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}naphthalene-2,7-disulfonic acid hydrate sodium
Synonyms: 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}naphthalene-2,7-disulfonic acid hydrate sodium
Molecular Weight (g/mol): 464.41
Molecular Formula: C17H15NNaO9S2
InChi Key: CYPHYAYBFNUBEV-UHFFFAOYSA-N
SMILES: O.[Na].OC1=CC(=CC2=CC(=CC(NC=C3C=CC=CC3=O)=C12)S(O)(=O)=O)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results