missing translation for 'onlineSavingsMsg'
Learn More

Azomethine H monosodium salt monohydrate, 95%

Numéro de catalogue. AC457090250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC457090250 25 g
AC457090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC457090250 Fournisseur Thermo Scientific Chemicals Code fournisseur 457090250
Il en reste null

CAS: 304655-82-1 | C17H15NNaO9S2 | 464.415 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 304655-82-1
Molecular Formula C17H15NNaO9S2
Molecular Weight (g/mol) 464.415
InChI Key CYPHYAYBFNUBEV-TTWKNDKESA-N
PubChem CID 131877441
IUPAC Name 4-hydroxy-5-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium;hydrate
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.O.[Na]

Spécifications

Infrared Spectrum Authentic
Formula Weight 463.41
Percent Purity 95%
Quantity 25 g
Chemical Name or Material Azomethine H monosodium salt monohydrate
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only