accessibility menu, dialog, popup

UserName

HOOSHSHOHO

CAS RN 304-55-2

IUPAC Name: 2,3-disulfanylbutanedioic acid
Synonyms: chemet succimer (substance) succimer, (R*,R*)-(+,-)- rhenium salt succimer (r*,r*)-(+,-)-succimer monosodium salt succimer succimer (product) (R*,R*)-(+,-)-isomer succimer serb brand of succimer dipotassium salt succimer
Molecular Weight (g/mol): 182.21
Molecular Formula: C4H6O4S2
InChi Key: ACTRVOBWPAIOHC-UHFFFAOYNA-N
SMILES: OC(=O)C(S)C(S)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (2)
  • (5)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)