accessibility menu, dialog, popup

UserName

CAS RN 3028-88-4

IUPAC Name: trioctyl phosphite
Synonyms: phosphorous acid, trioctyl ester
Molecular Weight (g/mol): 418.64
Molecular Formula: C24H51O3P
InChi Key: QOQNJVLFFRMJTQ-UHFFFAOYSA-N
SMILES: CCCCCCCCOP(OCCCCCCCC)OCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results