accessibility menu, dialog, popup

UserName

CAS RN 30186-18-6

IUPAC Name: 1-(4-bromo-2-hydroxyphenyl)ethan-1-one
Synonyms: acetophenone,2-hydroxy-4-bromo
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: LQCMMXGKEGWUIM-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)