missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. B53825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53825G 5 g
B53821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53825G Supplier TCI America Supplier No. B53825G

Chemical Identifiers

CAS 30186-18-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD03428533
InChI Key LQCMMXGKEGWUIM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
PubChem CID 10727403
IUPAC Name 1-(4-bromo-2-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=C(O)C=C(Br)C=C1

Specifications

Melting Point 42°C
Boiling Point 83°C
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-2′-hydroxyacetophenone