missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-hydroxyacetophenone, 95%

Catalog No. AAH6113706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6113706 5 g
AAH6113703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6113706 Supplier Thermo Scientific Chemicals Supplier No. H6113706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30186-18-6 | C8H7BrO2 | 215.05 g/mol

4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 30186-18-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD03428533
InChI Key LQCMMXGKEGWUIM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
PubChem CID 10727403
IUPAC Name 1-(4-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(Br)C=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 215.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4'-Bromo-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only