missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-methylimidazole-2-carboxylate, 95%, Thermo Scientific™

Catalog No. AAH6118514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6118514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6118514 Supplier Thermo Scientific Chemicals Supplier No. H6118514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 1-methylimidazole-2-carboxylate is used as a synthetic intermediate for solid phase synthesis of polyamides containing imidazole.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 30148-21-1
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD02683326
InChI Key NOTZYDYZBOBDFE-UHFFFAOYSA-N
Synonym ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester
PubChem CID 4261883
IUPAC Name ethyl 1-methylimidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1C

Specifications

Quantity 25 g
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol.
Formula Weight 154.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 1-methylimidazole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures