missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Methylimidazole-2-carboxylate 98.0+%, TCI America™

Catalog No. E11145G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11145G 5 g
E11141G 1 g
E111425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E11145G Supplier TCI America Supplier No. E11145G

Chemical Identifiers

CAS 30148-21-1
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD02683326
InChI Key NOTZYDYZBOBDFE-UHFFFAOYSA-N
Synonym ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester
PubChem CID 4261883
IUPAC Name ethyl 1-methyl-1H-imidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1C

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 1-Methylimidazole-2-carboxylate