accessibility menu, dialog, popup

UserName

CH3OFFFFFF

CAS RN 30071-93-3

IUPAC Name: 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[3,5-bis(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 256.15
Molecular Formula: C10H6F6O
InChi Key: MCYCSIKSZLARBD-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)