accessibility menu, dialog, popup

UserName

CAS RN 3002-48-0

IUPAC Name: tetrabutylazanium 4-nitrobenzen-1-olate
Synonyms: tetrabutylammonium ion p-nitrophenolate
Molecular Weight (g/mol): 380.57
Molecular Formula: C22H40N2O3
InChi Key: JWRHOHUAHHYNBL-UHFFFAOYSA-M
SMILES: [O-]C1=CC=C(C=C1)[N+]([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results