accessibility menu, dialog, popup

UserName

CAS RN 29882-07-3

CH3OClOH3C

CAS RN 29882-07-3

IUPAC Name: 4-chloro-1,1-dimethoxybutane
Synonyms: 4-chloro-1,1-dimethoxybutane
Molecular Weight (g/mol): 152.62
Molecular Formula: C6H13ClO2
InChi Key: LTLKJYMNUSSFAH-UHFFFAOYSA-N
SMILES: COC(CCCCl)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results