missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobutyraldehyde Dimethyl Acetal 97.0+%, TCI America™
$93.21 - $275.87
Chemical Identifiers
| CAS | 29882-07-3 |
|---|---|
| Molecular Formula | C6H13ClO2 |
| Molecular Weight (g/mol) | 152.618 |
| MDL Number | MFCD01632883 |
| InChI Key | LTLKJYMNUSSFAH-UHFFFAOYSA-N |
| Synonym | 4-Chloro-1,1-dimethoxybutane |
| PubChem CID | 122536 |
| IUPAC Name | 4-chloro-1,1-dimethoxybutane |
| SMILES | COC(CCCCl)OC |
Chemical Identifiers
| 29882-07-3 | |
| 152.618 | |
| LTLKJYMNUSSFAH-UHFFFAOYSA-N | |
| 122536 | |
| COC(CCCCl)OC |
| C6H13ClO2 | |
| MFCD01632883 | |
| 4-Chloro-1,1-dimethoxybutane | |
| 4-chloro-1,1-dimethoxybutane |
Specifications
| 29882-07-3 | |
| 120°C | |
| MFCD01632883 | |
| 1993 | |
| LTLKJYMNUSSFAH-UHFFFAOYSA-N | |
| 4-chloro-1,1-dimethoxybutane | |
| 122536 | |
| ≥97.0% (GC) | |
| 4-Chlorobutyraldehyde Dimethyl Acetal |
| Yellow | |
| C6H13ClO2 | |
| 5 g | |
| 4-Chloro-1,1-dimethoxybutane | |
| COC(CCCCl)OC | |
| 152.618 | |
| 152.62 | |
| Liquid |
Safety and Handling
TSCA : No