accessibility menu, dialog, popup

UserName

CH3H3CO

CAS RN 2979-19-3

IUPAC Name: 3,3-dimethylcyclohexan-1-one
Synonyms: cyclohexanone, 3,3-dimethyl-
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: ZVJQBBYAVPAFLX-UHFFFAOYSA-N
SMILES: CC1(C)CCCC(=O)C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)