missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylcyclohexanone, 97%, Thermo Scientific Chemicals

Catalog No. AAH2800006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2800006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2800006 Supplier Thermo Scientific Chemicals Supplier No. H2800006

An interest in the boll weevil sex attractants prompted an investigation into alternative syntheses of the starting material, 3, 3-dimethylcyclohexanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An interest in the boll weevil sex attractants prompted an investigation into alternative syntheses of the starting material, 3, 3-dimethylcyclohexanone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2979-19-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00040178
InChI Key ZVJQBBYAVPAFLX-UHFFFAOYSA-N
Synonym 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa
PubChem CID 76322
IUPAC Name 3,3-dimethylcyclohexan-1-one
SMILES CC1(C)CCCC(=O)C1

Specifications

Density 0.91
Boiling Point 175°C
Flash Point 59°C (138°F)
Refractive Index 1.447
Quantity 5 g
UN Number UN1224
Solubility Information Insoluble in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 3,3-Dimethylcyclohexanone
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only