accessibility menu, dialog, popup

UserName

CAS RN 29711-79-3

CH3NH3CNCS

CAS RN 29711-79-3

IUPAC Name: 4-isothiocyanato-N,N-dimethylnaphthalen-1-amine
Synonyms: 4-isothiocyanato-N,N-dimethylnaphthalen-1-amine
Molecular Weight (g/mol): 228.31
Molecular Formula: C13H12N2S
InChi Key: SMZHGTXTWIAKGS-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=CC2=C(C=C1)N=C=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results