accessibility menu, dialog, popup

UserName

H3COBrF

CAS RN 29578-39-0

IUPAC Name: 1-bromo-3-fluoro-5-methoxybenzene
Synonyms: 1-bromo-3-fluoro-5-methoxybenzene
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: XVNQVSGOIUYOPB-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results