accessibility menu, dialog, popup

UserName

CAS RN 2956-58-3

IUPAC Name: N-[2-(prop-2-enamido)ethyl]prop-2-enamide
Synonyms: N-[2-(prop-2-enamido)ethyl]prop-2-enamide
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: AYGYHGXUJBFUJU-UHFFFAOYSA-N
SMILES: C=CC(=O)NCCNC(=O)C=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)