missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Ethylenebisacrylamide, 96%

Catalog No. AAL1254406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1254406 5 g
AAL1254403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1254406 Supplier Thermo Scientific Chemicals Supplier No. L1254406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2956-58-3 | C8H12N2O2 | 168.196 g/mol

N,N'-Ethylenebisacrylamide is used in the preparation of N,N'-dicinnamoyl-ethylenediamine using poly(N-octadecylacrylamide) as a catalyst. It is also used as an intermediate in pharmaceutical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Ethylenebisacrylamide is used in the preparation of N,N′-dicinnamoyl-ethylenediamine using poly(N-octadecylacrylamide) as a catalyst. It is also used as an intermediate in pharmaceutical industries.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2956-58-3
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00042935
InChI Key AYGYHGXUJBFUJU-UHFFFAOYSA-N
Synonym n,n'-ethylenebisacrylamide, n,n-ethylenebisacrylamide, ethylene bis acrylamide, n,n'-ethane-1,2-diyl diacrylamide, 2-propenamide, n,n'-1,2-ethanediylbis, n,n'-bis acryloyl-1,2-diaminoethane, n,n'-bis acryloyl-1,2-diaminoethane, stabilized with hydroquinone, n-2-prop-2-enoylamino ethyl prop-2-enamide, n-2-prop-2-enamido ethyl prop-2-enamide, ethylenebisacrylamide
PubChem CID 168964
IUPAC Name N-[2-(prop-2-enoylamino)ethyl]prop-2-enamide
SMILES C=CC(=O)NCCNC(=O)C=C

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
Beilstein 1771570
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
Formula Weight 168.2
Percent Purity 96%
Chemical Name or Material N,N'-Ethylenebisacrylamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only