missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Ethylenebisacrylamide 97.0+%, TCI America™

Catalog No. E10865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10865G 5 g
E108625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10865G Supplier TCI America Supplier No. E10865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2956-58-3
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00042935
InChI Key AYGYHGXUJBFUJU-UHFFFAOYSA-N
Synonym n,n'-ethylenebisacrylamide, n,n-ethylenebisacrylamide, ethylene bis acrylamide, n,n'-ethane-1,2-diyl diacrylamide, 2-propenamide, n,n'-1,2-ethanediylbis, n,n'-bis acryloyl-1,2-diaminoethane, n,n'-bis acryloyl-1,2-diaminoethane, stabilized with hydroquinone, n-2-prop-2-enoylamino ethyl prop-2-enamide, n-2-prop-2-enamido ethyl prop-2-enamide, ethylenebisacrylamide
PubChem CID 168964
IUPAC Name N-[2-(prop-2-enoylamino)ethyl]prop-2-enamide
SMILES C=CC(=O)NCCNC(=O)C=C

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 5 g
Formula Weight 168.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Ethylenebisacrylamide
TSCA No
Recommended Storage Refrigerator