accessibility menu, dialog, popup

UserName

HOONCl

CAS RN 2942-59-8

IUPAC Name: 2-chloropyridine-3-carboxylic acid
Synonyms: 2-chloropyridine-3-carboxylic acid
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: IBRSSZOHCGUTHI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)