missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinic Acid 98.0+%, TCI America™

Catalog No. C110825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110825G 25 g
C1108250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C110825G Supplier TCI America Supplier No. C110825G

Chemical Identifiers

CAS 2942-59-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00006236
InChI Key IBRSSZOHCGUTHI-UHFFFAOYSA-N
Synonym 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
PubChem CID 76258
IUPAC Name 2-chloropyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Cl)C(=O)O

Specifications

Quantity 25 g
Formula Weight 157.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloronicotinic Acid
EINECSNumber (5)-6195
TSCA Yes