CAS RN 29331-92-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
10,11-Dihydro-10-hydroxycarbamazepine, ≥99% (HPLC), MilliporeSigma™ Supelco™
10,11-Dihydro-10-hydroxycarbamazepine is an active metabolite of the antiepileptic drug, oxcarbazepine.