missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-10-hydroxycarbamazepine, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111017967
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017967 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017967 Supplier Merck Emd Millipore Supplier No. 6134725MG

10,11-Dihydro-10-hydroxycarbamazepine is an active metabolite of the antiepileptic drug, oxcarbazepine.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 29331-92-8
Physical Form Neat
Linear Formula C15H14N2O2
MDL Number MFCD00871817
Synonym 10,11-Dihydro-10-hydroxy-5 H-dibenz[b,f]azepine-5-carboxamide; Licarbazepine
Type Analytical Standard
Quantity 25 mg
Formula Weight 254.28
Percent Purity ≥99% (HPLC)
Grade Analytical Standard