accessibility menu, dialog, popup

UserName

NBrClOOH

CAS RN 29241-65-4

IUPAC Name: 5-bromo-2-chloropyridine-3-carboxylic acid
Synonyms: 5-bromo-2-chloropyridine-3-carboxylic acid
Molecular Weight (g/mol): 236.45
Molecular Formula: C6H3BrClNO2
InChi Key: UKNYSJCAGUXDOQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CN=C1Cl
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)