missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloronicotinic Acid 98.0+%, TCI America™

Catalog No. B38835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38835G Supplier TCI America Supplier No. B38835G

Chemical Identifiers

CAS 29241-65-4
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.45
MDL Number MFCD03844847
InChI Key UKNYSJCAGUXDOQ-UHFFFAOYSA-N
Synonym 5-bromo-2-chloronicotinic acid, 5-bromo-2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo-2-chloro, 5-bromo-2-chloro-nicotinic acid, 5-bromo-3-carboxy-2-chloropyridine, 2-chloro-5-bromonicotinic acid, 5-bromo-2-chloronicotinicacid, pubchem12967, 5-bromo-2-chloro-pyridine-3-carboxylic acid, acmc-209h7p
PubChem CID 2763342
IUPAC Name 5-bromo-2-chloropyridine-3-carboxylic acid
SMILES OC(=O)C1=CC(Br)=CN=C1Cl

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 5 g
Formula Weight 236.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloronicotinic Acid