missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloronicotinic acid, 96%

Catalog No. AAH6115422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6115422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6115422 Supplier Thermo Scientific Chemicals Supplier No. H6115422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29241-65-4 | C6H3BrClNO2 | 236.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29241-65-4
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.45
MDL Number MFCD03844847
InChI Key UKNYSJCAGUXDOQ-UHFFFAOYSA-N
Synonym 5-bromo-2-chloronicotinic acid, 5-bromo-2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo-2-chloro, 5-bromo-2-chloro-nicotinic acid, 5-bromo-3-carboxy-2-chloropyridine, 2-chloro-5-bromonicotinic acid, 5-bromo-2-chloronicotinicacid, pubchem12967, 5-bromo-2-chloro-pyridine-3-carboxylic acid, acmc-209h7p
PubChem CID 2763342
IUPAC Name 5-bromo-2-chloropyridine-3-carboxylic acid
SMILES OC(=O)C1=CC(Br)=CN=C1Cl

Specifications

Quantity 100 g
Formula Weight 236.45
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chloronicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only