accessibility menu, dialog, popup

UserName

ClSOOBr

CAS RN 2905-24-0

IUPAC Name: 3-bromobenzene-1-sulfonyl chloride
Synonyms: 3-bromobenzenesulfonyl chloride
Molecular Weight (g/mol): 255.51
Molecular Formula: C6H4BrClO2S
InChi Key: PJGOLCXVWIYXRQ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)