missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzenesulfonyl chloride, 97%

Catalog No. AAA1150714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1150714 25 g
AAA1150703 1 g
AAA1150706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1150714 Supplier Thermo Scientific Chemicals Supplier No. A1150714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2905-24-0 | C6H4BrClO2S | 255.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-24-0
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00052313
InChI Key PJGOLCXVWIYXRQ-UHFFFAOYSA-N
Synonym 3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
PubChem CID 17943
IUPAC Name 3-bromobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC(Br)=C1

Specifications

Melting Point 30°C to 33°C
Density 1.773
Boiling Point 90°C to 91°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.593
Quantity 25 g
UN Number UN3261
Beilstein 2691656
Sensitivity Moisture sensitive
Formula Weight 255.52
Percent Purity 97%
Chemical Name or Material 3-Bromobenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only