accessibility menu, dialog, popup

UserName

CAS RN 289039-29-8

INCl

CAS RN 289039-29-8

IUPAC Name: 2-chloro-5-iodobenzonitrile
Synonyms: 2-chloro-5-iodobenzonitrile
Molecular Weight (g/mol): 263.46
Molecular Formula: C7H3ClIN
InChi Key: RXNOBNYCCAEGJD-UHFFFAOYSA-N
SMILES: ClC1=C(C=C(I)C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results