accessibility menu, dialog, popup

UserName

SN

CAS RN 288-16-4

IUPAC Name: 1,2-thiazole
Synonyms: isothiazole
Molecular Weight (g/mol): 85.12
Molecular Formula: C3H3NS
InChi Key: ZLTPDFXIESTBQG-UHFFFAOYSA-N
SMILES: S1C=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)