missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isothiazole, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Flash Point | 22°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Quantity | 5 g |
| Formula Weight | 85.13 |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Chemical Name or Material | Isothiazole |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wash face, hands and any exposed skin thoroughly after handling. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. |
RUO – Research Use Only