accessibility menu, dialog, popup

UserName

CAS RN 28342-75-8

FBrBrF

CAS RN 28342-75-8

IUPAC Name: 1,5-dibromo-2,4-difluorobenzene
Synonyms: 1,5-dibromo-2,4-difluorobenzene
Molecular Weight (g/mol): 271.89
Molecular Formula: C6H2Br2F2
InChi Key: PPUZKAPOPPRMFE-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(Br)C=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results