missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-2,4-difluorobenzene 98.0+%, TCI America™

Catalog No. D52591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D52591G Supplier TCI America Supplier No. D52591G
Only null left

Chemical Identifiers

CAS 28342-75-8
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.89
MDL Number MFCD08458122
InChI Key PPUZKAPOPPRMFE-UHFFFAOYSA-N
PubChem CID 10564511
IUPAC Name 1,5-dibromo-2,4-difluorobenzene
SMILES FC1=CC(F)=C(Br)C=C1Br

Specifications

Melting Point 31°C
Color White-Yellow
Boiling Point 102°C
Quantity 1 g
Formula Weight 271.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,5-Dibromo-2,4-difluorobenzene
TSCA No
Recommended Storage Refrigerator