accessibility menu, dialog, popup

UserName

CAS RN 28320-32-3

IUPAC Name: 2,7-dibromo-9,9-dimethyl-9H-fluorene
Synonyms: 2,7-dibromo-9,9-dimethylfluorene
Molecular Weight (g/mol): 352.07
Molecular Formula: C15H12Br2
InChi Key: LONBOJIXBFUBKQ-UHFFFAOYSA-N
SMILES: CC1(C)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results