accessibility menu, dialog, popup

UserName

CAS RN 28308-00-1

H3CNCH3NCH3CH3ClZnCl

CAS RN 28308-00-1

IUPAC Name: [2-(dimethylamino)ethyl]dimethylamine; dichlorozinc
Synonyms: temed; zinc chloride
Molecular Weight (g/mol): 252.49
Molecular Formula: C6H16Cl2N2Zn
InChi Key: WEHCCWCYFYMBQX-UHFFFAOYSA-L
SMILES: Cl[Zn]Cl.CN(C)CCN(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results