missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(N,N,N',N'-tetramethylethylenediamine)zinc(II) 98.0+%, TCI America™

Catalog No. D439325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D439325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D439325G Supplier TCI America Supplier No. D439325G
Only null left

Chemical Identifiers

CAS 28308-00-1
Molecular Formula C6H16Cl2N2Zn
Molecular Weight (g/mol) 252.488
MDL Number MFCD00013233
InChI Key WEHCCWCYFYMBQX-UHFFFAOYSA-L
Synonym Dichloro(N,N,N′C,N′C-tetramethylethane-1,2-diamine)zinc(II), (N,N,N′C,N′C-Tetramethylethylenediamine)zinc(II) Dichloride, ZnCl2-TMEDA
PubChem CID 6096193
IUPAC Name dichlorozinc;N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C.Cl[Zn]Cl

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 25 g
UN Number 3263
Formula Weight 252.49
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloro(N,N,N′,N′-tetramethylethylenediamine)zinc(II)