accessibility menu, dialog, popup

UserName

CAS RN 2802-68-8

IUPAC Name: bis(methanol); trifluoroborane
Synonyms: boron trifluoride; bis(methanol)
Molecular Weight (g/mol): 131.89
Molecular Formula: C2H8BF3O2
InChi Key: MPRMFRXCVMCOMX-UHFFFAOYSA-N
SMILES: CO.CO.FB(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)