missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride dimethanol complex, 50-52 wt% BF3

Catalog No. AC158902500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158902500 250 g Glass bottle
AC158900500 50 g Glass bottle
AC158900010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158902500 Supplier Thermo Scientific Chemicals Supplier No. 158902500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2802-68-8 | C2H8BF3O2 | 131.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 2802-68-8 , 7637-07-2 , 67-56-1
Molecular Formula C2H8BF3O2
Molecular Weight (g/mol) 131.89
MDL Number MFCD00071635
InChI Key MPRMFRXCVMCOMX-UHFFFAOYSA-N
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
PubChem CID 11062313
SMILES CO.CO.FB(F)F

Specifications

Melting Point -20.0°C
Color Colorless to Yellow
Density 1.2220g/mL
Boiling Point 59.0°C (4.0 mmHg)
Flash Point 68°C
Packaging Glass bottle
Refractive Index 1.34
Linear Formula BF3·2CH3OH
Quantity 250 g
Specific Gravity 1.222
Solubility Information Solubility in water: hydrolysis.
IUPAC Name bis(methanol); trifluoroborane
Formula Weight 131.89
Percent Purity ≥50% (BF3)
Physical Form Liquid
Chemical Name or Material Boron trifluoride dimethanol complex, 50-52 wt% BF3
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.<br/
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-543-9
TSCA TSCA

RUO – Research Use Only