missing translation for 'onlineSavingsMsg'
Learn More

Boron Trifluoride - Methanol Complex, TCI America™

Catalog No. B0894500ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0894500ML 500 mL
B0894100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0894500ML Supplier TCI America Supplier No. B0894500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2802-68-8
Molecular Formula C2H8BF3O2
Molecular Weight (g/mol) 131.89
MDL Number MFCD00071635
InChI Key MPRMFRXCVMCOMX-UHFFFAOYSA-N
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
PubChem CID 11062313
IUPAC Name bis(methanol); trifluoroborane
SMILES CO.CO.FB(F)F

Specifications

Color Yellow
Quantity 500 mL
UN Number 2920
Formula Weight 131.89
Physical Form Liquid
Chemical Name or Material Boron Trifluoride - Methanol Complex
EINECSNumber (2)-2112
RTECSNumber ED2275000
TSCA Yes