accessibility menu, dialog, popup

UserName

CH3OOFFFFFFFF

CAS RN 2795-50-8

IUPAC Name: ethyl 2,2,3,3,4,4,5,5-octafluoropentanoate
Synonyms: ethyl 2,2,3,3,4,4,5,5-octafluoropentanoate
Molecular Weight (g/mol): 274.11
Molecular Formula: C7H6F8O2
InChi Key: HXWMNJVBQLBDGW-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results