missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5H-Octafluorovalerate 98.0+%, TCI America™

Catalog No. E10225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10225G 5 g
E102225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10225G Supplier TCI America Supplier No. E10225G
Only null left

Chemical Identifiers

CAS 2795-50-8
Molecular Formula C7H6F8O2
Molecular Weight (g/mol) 274.11
MDL Number MFCD00153153
InChI Key HXWMNJVBQLBDGW-UHFFFAOYSA-N
Synonym 5H-Octafluorovaleric Acid Ethyl Ester
PubChem CID 2737223
IUPAC Name ethyl 2,2,3,3,4,4,5,5-octafluoropentanoate
SMILES CCOC(=O)C(C(C(C(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 180°C
Quantity 5 g
UN Number 3272
Formula Weight 274.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 5H-Octafluorovalerate