CAS RN 27896-84-0
CAS RN 27896-84-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Nitrobenzimidazole Nitrate 98.0+%, TCI America™
CAS: 27896-84-0 Molecular Formula: C7H6N4O5 Molecular Weight (g/mol): 226.148 MDL Number: MFCD00013157 InChI Key: ZUZQXHSOEZUAIS-UHFFFAOYSA-N PubChem CID: 119743 IUPAC Name: nitric acid;6-nitro-1H-benzimidazole SMILES: C1=CC2=C(C=C1[N+](=O)[O-])NC=N2.[N+](=O)(O)[O-]