missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzimidazole Nitrate 98.0+%, TCI America™

Catalog No. p-7133648
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015125G 25 g
N0151250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N015125G Supplier TCI America Supplier No. N015125G
Only null left

Chemical Identifiers

CAS 27896-84-0
Molecular Formula C7H6N4O5
Molecular Weight (g/mol) 226.148
MDL Number MFCD00013157
InChI Key ZUZQXHSOEZUAIS-UHFFFAOYSA-N
PubChem CID 119743
IUPAC Name nitric acid;6-nitro-1H-benzimidazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])NC=N2.[N+](=O)(O)[O-]

Specifications

Melting Point 222°C
Color Yellow
Quantity 25 g
Formula Weight 226.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrobenzimidazole Nitrate
RTECSNumber DD9801000
TSCA Yes