missing translation for 'onlineSavingsMsg'
Learn More

Nicergoline, ≥97% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111013805
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013805 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013805 Supplier Merck Emd Millipore Supplier No. N7889100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nicergoline is an ergot derivative, which may be used in the treatment of affective, cognitive and behavioral disorders seen in older people. It is an α -adrenergic antagonist that may be used for companion animals like cats and dogs to treat, behavioral disorders, problems with aging, excessive vocaliztion and restessness.

Specifications

CAS 27848-84-6
Physical Form Neat
Linear Formula C24H26 BrN3O3
MDL Number MFCD00869626
Synonym 5-Bromonicotinic acid 10-methoxy-1,6-dimethylergoline-8-methyl ester
Type Analytical Standard
Industry Type Drug Analysis
Quantity 100 mg
Formula Weight 484.39
Percent Purity ≥97% (TLC)
Grade Analytical Standard
Show More Show Less