accessibility menu, dialog, popup

UserName

H3C(S)NH2OOCH3CH3HCl

CAS RN 2743-40-0

IUPAC Name: ethyl (2S)-2-amino-4-methylpentanoate
Synonyms: leucine, ethyl ester
Molecular Weight (g/mol): 159.23
Molecular Formula: C8H17NO2
InChi Key: QIGLJVBIRIXQRN-ZETCQYMHSA-N
SMILES: CCOC(=O)[C@@H](N)CC(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)