accessibility menu, dialog, popup

UserName

CAS RN 273-09-6

IUPAC Name: 2,1,3-benzoxadiazole
Synonyms: benzofurazan
Molecular Weight (g/mol): 120.11
Molecular Formula: C6H4N2O
InChi Key: AWBOSXFRPFZLOP-UHFFFAOYSA-N
SMILES: O1N=C2C=CC=CC2=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)