missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzoxadiazole 98.0+%, TCI America™

Catalog No. B44735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44735G 5 g
B44731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44735G Supplier TCI America Supplier No. B44735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 273-09-6
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
MDL Number MFCD00068064
InChI Key AWBOSXFRPFZLOP-UHFFFAOYSA-N
Synonym benzofurazan, benzo c 1,2,5 oxadiazole, benzofurazane, 2,1,3-benzoxadiazol, benzo-2,1,3-oxadiazole, 2-oxa-1,3-diaza-2h-isoindene, 3,5-cyclohexadiene-1,2-dione, dioxime, furazan, pubchem22999, acmc-209gwb
PubChem CID 67501
IUPAC Name 2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C=C1

Specifications

Melting Point 53°C
Color Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 120.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,1,3-Benzoxadiazole
RTECSNumber DM2580000
TSCA No