CAS RN 2670-38-4
IUPAC Name:
3,4-dichlorobenzamide
Synonyms:
3,4-dichlorobenzamide
Molecular Weight (g/mol):
190.02
Molecular Formula:
C7H5Cl2NO
InChi Key:
XURBWYCGQQXTHJ-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
3,4-Dichlorobenzamide 98.0+%, TCI America™
CAS: 2670-38-4 Molecular Formula: C7H5Cl2NO Molecular Weight (g/mol): 190.02 MDL Number: MFCD00053009 InChI Key: XURBWYCGQQXTHJ-UHFFFAOYSA-N Synonym: benzamide, 3,4-dichloro,acmc-20anfl,pubchem3462,3,4-dichloro benzamide,benzamide,3,4-dichloro,3,4-bis chloranyl benzamide PubChem CID: 137671 IUPAC Name: 3,4-dichlorobenzamide SMILES: NC(=O)C1=CC=C(Cl)C(Cl)=C1