missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-tert-butylbenzoate, 99%, Thermo Scientific™

Catalog No. AC173480250
Change view
Click to view available options
Quantity:
25g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173480250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173480250 Supplier Thermo Scientific Chemicals Supplier No. 173480250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26537-19-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.26
MDL Number MFCD00008835
InChI Key UPIJOAFHOIWPLT-UHFFFAOYSA-N
Synonym methyl 4-tert-butyl benzoate, methyl p-tert-butylbenzoate, 4-tert-butylbenzoic acid methyl ester, unii-8vk4o27jmt, benzoic acid, 4-1,1-dimethylethyl-, methyl ester, methyl 4-t-butylbenzoate, 8vk4o27jmt, methyl4-tert-butylbenzoate, methyl p-t-butylbenzoate, benzoic acid, p-tert-butyl-, methyl ester
PubChem CID 97433
IUPAC Name methyl 4-tert-butylbenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OC

Specifications

Density 0.9900g/mL
Boiling Point 122.0°C to 124.0°C (9.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5090 to 1.5110
Linear Formula (CH3)3CC6H4CO2CH3
Quantity 25g
Beilstein 09, 560
Specific Gravity 0.99
Formula Weight 192.26
Percent Purity 99%
Chemical Name or Material Methyl 4-tert-butylbenzoate, 99%
Show More Show Less
EINECSNumber 247-768-5

RUO – Research Use Only